3-Chloro-4-methyl-2-(methylthio)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

32534-500MG 32534-1G 32534-5G 32534-25G
77325-354EA 83.43 USD
77325-354 77342-178 77361-718 77373-352
3-Chloro-4-methyl-2-(methylthio)phenylboronic acid ≥97%
3-Chloro-4-methyl-2-(methylthio)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BClO₂S
MW: 216.493 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529820
CAS Number: 2121513-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR