3-Hydroxy-4-iodopyridine HCl ≥95%

Supplier: AOBChem USA

Warning

32533-1G 32533-5G 32533-25G
77340-872EA 61.8 USD
77340-872 77361-716 77373-350
3-Hydroxy-4-iodopyridine HCl ≥95%
3-Hydroxy-4-iodopyridine HCl

Precaución: for Research Use Only

Formula: C₅H₅ClINO
MW: 257.457 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529819
CAS Number: 2056110-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR