2-Fluoro-4-methoxyphenyl acetate ≥97%

Supplier: AOBChem USA

Warning

32527-500MG 32527-1G 32527-5G 32527-25G
77325-352EA 128.75 USD
77325-352 77344-236 77361-714 77373-348
2-Fluoro-4-methoxyphenyl acetate ≥97%
2-Fluoro-4-methoxyphenyl acetate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD28364548
CAS Number: 181305-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR