4-Bromo-2,3-difluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32511-1G 32511-5G 32511-25G
77340-636EA 60.77 USD
77340-636 77361-712 77373-346
4-Bromo-2,3-difluorophenylboronic acid pinacol ester ≥97%
4-Bromo-2,3-difluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₄BBrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28142175
CAS Number: 2121515-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR