2-Chloro-5-fluoro-3-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

32498-500MG 32498-1G 32498-5G 32498-25G
77325-350EA 243.08 USD
77325-350 77349-092 77361-710 77384-828
2-Chloro-5-fluoro-3-(trifluoromethyl)phenol ≥97%
2-Chloro-5-fluoro-3-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₃ClF₄O
MW: 214.545 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529816
CAS Number: 2056110-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR