2-Chloro-5-fluoro-3-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

32495-500MG 32495-1G 32495-5G 32495-25G
77325-348EA 259.56 USD
77325-348 77349-090 77361-708 77384-826
2-Chloro-5-fluoro-3-(trifluoromethyl)benzyl alcohol ≥97%
2-Chloro-5-fluoro-3-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₅ClF₄O
MW: 228.571 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529813
CAS Number: 2056110-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR