3-Iodo-4-methylthiophene ≥95%

Supplier: AOBChem USA

Warning

32471-500MG 32471-1G 32471-5G 32471-25G
77325-346EA 140.08 USD
77325-346 77344-700 77361-706 77384-824
3-Iodo-4-methylthiophene ≥95%
3-Iodo-4-methylthiophene

Precaución: for Research Use Only

Formula: C₅H₅IS
Storage Temperature: Ambient
MDL Number: MFCD18417231
CAS Number: 16494-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR