6-Fluoro-2-methoxy-3-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32460-500MG 32460-1G 32460-5G 32460-25G
77325-344EA 219.39 USD
77325-344 77349-088 77361-704 77384-822
6-Fluoro-2-methoxy-3-methylphenylboronic acid pinacol ester ≥97%
6-Fluoro-2-methoxy-3-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BFO₃
MW: 266.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22415000
CAS Number: 2121513-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR