1-(Benzyloxy)-2-chloro-4,5-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

32423-500MG 32423-1G 32423-5G
77336-336EA 387.28 USD
77336-336 77349-084 77373-340
1-(Benzyloxy)-2-chloro-4,5-difluorobenzene ≥95%
1-(Benzyloxy)-2-chloro-4,5-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD33403711
CAS Number: 2624417-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR