Ethyl 5-chloro-2,4-dimethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

32336-500MG 32336-1G 32336-5G 32336-25G
77325-336EA 202.91 USD
77325-336 77349-080 77361-696 77384-816
Ethyl 5-chloro-2,4-dimethoxybenzoate ≥97%
Ethyl 5-chloro-2,4-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₄
Storage Temperature: Ambient
MDL Number: MFCD24108518
CAS Number: 1022091-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR