Methyl 3-methoxy-4-(trifluoromethoxy)benzoate ≥97%

Supplier: AOBChem USA

Warning

32324-500MG 32324-1G 32324-5G
77339-470EA 593.28 USD
77339-470 77349-076 77373-332
Methyl 3-methoxy-4-(trifluoromethoxy)benzoate ≥97%
Methyl 3-methoxy-4-(trifluoromethoxy)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD18399559
CAS Number: 1261827-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR