3-Mthoxy-4-(trifluoromethoxy)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32318-500MG 32318-1G 32318-5G 32318-25G
77325-334EA 201.88 USD
77325-334 77349-074 77361-694 77384-814
3-Mthoxy-4-(trifluoromethoxy)phenylboronic acid pinacol ester ≥97%
3-Mthoxy-4-(trifluoromethoxy)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BF₃O₄
Storage Temperature: Ambient
MDL Number: MFCD18760110
CAS Number: 1623766-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR