Methyl 3-chloro-5-fluoro-4-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

32291-500MG 32291-1G 32291-5G 32291-25G
77325-332EA 260.59 USD
77325-332 77349-072 77361-692 77384-812
Methyl 3-chloro-5-fluoro-4-methoxybenzoate ≥97%
Methyl 3-chloro-5-fluoro-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₃
MW: 218.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24108541
CAS Number: 1520844-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR