3-Ethyl-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32268-500MG 32268-1G 32268-5G
77325-330EA 336.81 USD
77325-330 77349-068 77373-328
3-Ethyl-4-methoxybenzoic acid ≥97%
3-Ethyl-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14538394
CAS Number: 22934-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR