1-(5-Bromopyridin-2-yl)-4-oxocyclohexan-1-ol ≥97%

Supplier: AOBChem USA

Warning

32151-500MG 32151-1G 32151-5G 32151-25G
77325-324EA 193.64 USD
77325-324 77349-062 77361-684 77384-806
1-(5-Bromopyridin-2-yl)-4-oxocyclohexan-1-ol ≥97%
1-(5-Bromopyridin-2-yl)-4-oxocyclohexan-1-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD31619439
CAS Number: 857650-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR