3,4-Dimethoxy-2-(isobutoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

32118-500MG 32118-1G 32118-5G
77336-234EA 384.19 USD
77336-234 77349-060 77373-320
3,4-Dimethoxy-2-(isobutoxy)phenylboronic acid ≥97%
3,4-Dimethoxy-2-(isobutoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₉BO₅
MW: 254.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29086467
CAS Number: 1345712-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR