1-Bromo-2-isobutoxy-3,4-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

32117-500MG 32117-1G 32117-5G 32117-25G
77325-322EA 214.24 USD
77325-322 77349-058 77361-680 77384-804
1-Bromo-2-isobutoxy-3,4-dimethoxybenzene ≥97%
1-Bromo-2-isobutoxy-3,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₇BrO₃
Storage Temperature: Ambient
MDL Number: MFCD29086468
CAS Number: 1345712-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR