2-Chloro-4-(methoxymethoxy)-benzoic acid ≥97%

Supplier: AOBChem USA

Warning

32098-1G 32098-5G 32098-25G
77340-976EA 65.92 USD
77340-976 77361-678 77373-318
2-Chloro-4-(methoxymethoxy)-benzoic acid ≥97%
2-Chloro-4-(methoxymethoxy)-benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₄
Storage Temperature: Ambient
MDL Number: MFCD31619437
CAS Number: 1700623-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR