2-Chloro-3-fluoro-4-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

32040-500MG 32040-1G 32040-5G
77339-314EA 577.83 USD
77339-314 77349-054 77373-316
2-Chloro-3-fluoro-4-formylphenylboronic acid ≥97%
2-Chloro-3-fluoro-4-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClFO₃
MW: 202.375 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30344681
CAS Number: 2121514-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR