Ethyl 6-chloro-5-methoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

32025-500MG 32025-1G 32025-5G 32025-25G
77325-318EA 161.71 USD
77325-318 77345-834 77361-674 77384-800
Ethyl 6-chloro-5-methoxynicotinate ≥97%
Ethyl 6-chloro-5-methoxynicotinate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD18257298
CAS Number: 1256821-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR