Benzyl 6-bromo-5-methoxypyridine-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

32020-500MG 32020-1G 32020-5G 32020-25G
77325-316EA 263.68 USD
77325-316 77349-052 77361-672 77384-798
Benzyl 6-bromo-5-methoxypyridine-3-carboxylate ≥97%
Benzyl 6-bromo-5-methoxypyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrNO₃
MW: 322.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30344678
CAS Number: 1993479-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR