Ethyl 6-bromo-5-methoxypyridine-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

32019-500MG 32019-1G 32019-5G 32019-25G
77325-314EA 284.28 USD
77325-314 77349-050 77361-670 77384-796
Ethyl 6-bromo-5-methoxypyridine-3-carboxylate ≥97%
Ethyl 6-bromo-5-methoxypyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₃
MW: 260.085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28741717
CAS Number: 1807164-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR