4-Bromo-1-methoxy-2-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

31998-500MG 31998-1G 31998-5G 31998-25G
77325-312EA 85.49 USD
77325-312 77342-240 77361-668 77373-312
4-Bromo-1-methoxy-2-(methoxymethyl)benzene ≥97%
4-Bromo-1-methoxy-2-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD14686969
CAS Number: 338454-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR