3-Bromo-4-fluoro-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

31796-500MG 31796-1G 31796-5G
77337-120EA 408.91 USD
77337-120 77349-040 77373-298
3-Bromo-4-fluoro-2-methoxybenzaldehyde ≥97%
3-Bromo-4-fluoro-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28967915
CAS Number: 1784176-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR