Benzyl 5-bromo-2-fluoropyridine-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

31756-500MG 31756-1G 31756-5G
77338-688EA 510.88 USD
77338-688 77349-034 77373-292
Benzyl 5-bromo-2-fluoropyridine-4-carboxylate ≥97%
Benzyl 5-bromo-2-fluoropyridine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₉BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD30344674
CAS Number: 1993479-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR