2,6-Difluoro-4-isopropoxybenzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

31752-500MG 31752-1G 31752-5G 31752-25G
77325-302EA 243.08 USD
77325-302 77349-032 77361-658 77384-788
2,6-Difluoro-4-isopropoxybenzoic acid methyl ester ≥97%
2,6-Difluoro-4-isopropoxybenzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₃
MW: 230.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23114999
CAS Number: 1394944-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR