2,6-Difluoro-4-isopropyloxybenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

31750-500MG 31750-1G 31750-5G 31750-25G
77325-300EA 140.08 USD
77325-300 77344-696 77361-656 77384-786
2,6-Difluoro-4-isopropyloxybenzyl alcohol ≥95%
2,6-Difluoro-4-isopropyloxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂O₂
MW: 202.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27950790
CAS Number: 438049-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR