2-Ethoxy-4-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

31741-500MG 31741-1G 31741-5G 31741-25G
77325-294EA 244.11 USD
77325-294 77349-026 77361-650 77384-780
2-Ethoxy-4-(trifluoromethoxy)benzoic acid ≥97%
2-Ethoxy-4-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₄
MW: 250.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27933466
CAS Number: 933063-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR