2-Ethoxy-4-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

31739-500MG 31739-1G 31739-5G
77325-292EA 342.99 USD
77325-292 77349-024 77373-288
2-Ethoxy-4-(trifluoromethoxy)benzaldehyde ≥97%
2-Ethoxy-4-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₃
MW: 234.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755901
CAS Number: 1071156-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR