4-Benzyloxy-2,6-difluorobenzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

31733-500MG 31733-1G 31733-5G
77335-666EA 363.59 USD
77335-666 77349-022 77373-286
4-Benzyloxy-2,6-difluorobenzoic acid methyl ester ≥97%
4-Benzyloxy-2,6-difluorobenzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₅H₁₂F₂O₃
MW: 278.251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30344671
CAS Number: 1993479-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR