Methyl 2,4-difluoro-5-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

31730-500MG 31730-1G 31730-25G 31730-5G
77325-286EA 286.34 USD
77325-286 77349-020 77384-774 77373-284
Methyl 2,4-difluoro-5-methoxybenzoate ≥97%
Methyl 2,4-difluoro-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28789274
CAS Number: 1804416-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR