1-Bromo-2-fluoro-3,4-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

31648-500MG 31648-1G 31648-5G
77338-546EA 498.52 USD
77338-546 77349-018 77373-280
1-Bromo-2-fluoro-3,4-dimethoxybenzene ≥95%
1-Bromo-2-fluoro-3,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28401603
CAS Number: 1095544-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR