Ethyl 2-bromo-4,6-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

31623-500MG 31623-1G 31623-5G
77337-046EA 407.88 USD
77337-046 77349-016 77373-276
Ethyl 2-bromo-4,6-difluorobenzoate ≥97%
Ethyl 2-bromo-4,6-difluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrF₂O₂
MW: 265.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28737897
CAS Number: 1807243-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR