2,4-Dichloro-3-methoxyphenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
31595-500MG
31595-1G
31595-5G
31595-25G
77325-282EA
123.6
USD
77325-282
77344-030
77361-640
77373-274
2,4-Dichloro-3-methoxyphenylboronic acid pinacol ester ≥97%
2,4-Dichloro-3-methoxyphenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₇BCl₂O₃ MW: 302.989 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29915594 CAS Number: 1689538-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...