5-(Difluoromethoxy)picolinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-(Difluoromethoxy)pyridine-2-carboxylic acid

31572-500MG 31572-1G 31572-5G 31572-25G
77325-278EA 220.42 USD
77325-278 77349-010 77361-636 77384-770
5-(Difluoromethoxy)picolinic acid ≥97%
5-(Difluoromethoxy)picolinic acid

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₃
MW: 189.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18261162
CAS Number: 1174323-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR