5-(Cyclopropylmethoxy)pyridine-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

31570-500MG 31570-1G 31570-5G 31570-25G
77325-276EA 252.35 USD
77325-276 77349-008 77361-634 77384-768
5-(Cyclopropylmethoxy)pyridine-2-carboxylic acid ≥97%
5-(Cyclopropylmethoxy)pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₃
MW: 193.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20646229
CAS Number: 1266787-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR