3,5-Dimethyl-2-methoxyphenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

31516-500MG 31516-1G 31516-25G 31516-5G
77325-272EA 304.88 USD
77325-272 77349-004 77384-766 77373-268
3,5-Dimethyl-2-methoxyphenylboronic acid pinacol ester ≥95%
3,5-Dimethyl-2-methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₃BO₃
MW: 262.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414995
CAS Number: 2121512-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR