3,5-Difluoro-4-(methylthio)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31515-500MG 31515-1G 31515-5G
77337-836EA 449.08 USD
77337-836 77349-002 77373-266
3,5-Difluoro-4-(methylthio)phenylboronic acid pinacol ester ≥97%
3,5-Difluoro-4-(methylthio)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD29915591
CAS Number: 2121514-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR