1-Chloro-2,3-dimethoxy-4-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

31513-500MG 31513-1G 31513-5G 31513-25G
77325-270EA 204.97 USD
77325-270 77349-000 77361-632 77384-764
1-Chloro-2,3-dimethoxy-4-fluorobenzene ≥97%
1-Chloro-2,3-dimethoxy-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈ClFO₂
MW: 190.599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751432
CAS Number: 1804409-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR