2,5-Difluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

31506-500MG 31506-1G 31506-5G 31506-25G
77325-266EA 252.35 USD
77325-266 77348-996 77361-628 77384-760
2,5-Difluoro-4-methylbenzaldehyde ≥97%
2,5-Difluoro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD16250131
CAS Number: 879093-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR