4-Chloro-2,6-dimethyl-benzoic acid ethyl ester ≥95%

Supplier: AOBChem USA

Warning

31496-1G 31496-5G 31496-25G
77341-730EA 103 USD
77341-730 77361-626 77373-264
4-Chloro-2,6-dimethyl-benzoic acid ethyl ester ≥95%
4-Chloro-2,6-dimethyl-benzoic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD11042977
CAS Number: 1243410-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR