Methyl 4-chloro-2,6-dimethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

31495-500MG 31495-1G 31495-5G 31495-25G
77325-264EA 160.68 USD
77325-264 77345-770 77361-624 77384-758
Methyl 4-chloro-2,6-dimethylbenzoate ≥95%
Methyl 4-chloro-2,6-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD29915590
CAS Number: 352278-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR