3-(Benzyloxy)-6-bromo-2-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31486-1G 31486-5G 31486-25G
77341-396EA 84.46 USD
77341-396 77361-622 77373-262
3-(Benzyloxy)-6-bromo-2-fluorophenylboronic acid pinacol ester ≥97%
3-(Benzyloxy)-6-bromo-2-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₁BBrFO₃
Storage Temperature: Ambient
MDL Number: MFCD29915589
CAS Number: 2121514-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR