3-Ethoxy-5-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

31448-500MG 31448-1G 31448-5G 31448-25G
77325-260EA 181.28 USD
77325-260 77346-808 77361-618 77384-754
3-Ethoxy-5-(trifluoromethyl)phenol ≥97%
3-Ethoxy-5-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₉H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD29044607
CAS Number: 1881320-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR