[2-Methoxy-6-(methylsulfanyl)phenyl]methanol ≥97%

Supplier: AOBChem USA

Warning

31375-1G 31375-5G 31375-25G
77340-948EA 64.89 USD
77340-948 77361-608 77373-260
[2-Methoxy-6-(methylsulfanyl)phenyl]methanol ≥97%
[2-Methoxy-6-(methylsulfanyl)phenyl]methanol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD18905253
CAS Number: 1379034-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR