3-Chloro-2,4-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

31336-500MG 31336-1G 31336-5G 31336-25G
77325-248EA 187.46 USD
77325-248 77348-988 77361-604 77384-742
3-Chloro-2,4-dimethylbenzoic acid ≥97%
3-Chloro-2,4-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD30529804
CAS Number: 1632385-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR