(2-Chloro-6-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

31331-500MG 31331-1G 31331-5G 31331-25G
77325-246EA 243.08 USD
77325-246 77348-986 77361-602 77384-740
(2-Chloro-6-iodophenyl)methanol ≥97%
(2-Chloro-6-iodophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₆ClIO
Storage Temperature: Ambient
MDL Number: MFCD18072887
CAS Number: 945543-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR