Ethyl 4-methoxy-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%

Supplier: AOBChem USA

Warning

31328-500MG 31328-1G 31328-5G 31328-25G
77325-244EA 147.29 USD
77325-244 77345-196 77361-600 77384-738
Ethyl 4-methoxy-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%
Ethyl 4-methoxy-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₆H₂₃BO₅
Storage Temperature: Ambient
MDL Number: MFCD22494079
CAS Number: 214360-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR