2-(4-(Ethoxymethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%

Supplier: AOBChem USA

Warning

31281-500MG 31281-1G 31281-5G 31281-25G
77325-242EA 134.93 USD
77325-242 77344-434 77361-596 77384-736
2-(4-(Ethoxymethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
2-(4-(Ethoxymethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₅H₂₃BO₃
MW: 262.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22493780
CAS Number: 1351353-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR