Ethyl 4-(benzyloxy)-2,3-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

31277-500MG 31277-1G 31277-5G 31277-25G
77325-240EA 284.28 USD
77325-240 77348-984 77361-594 77384-734
Ethyl 4-(benzyloxy)-2,3-difluorobenzoate ≥97%
Ethyl 4-(benzyloxy)-2,3-difluorobenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₄F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD29088286
CAS Number: 1879026-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR